Invitrogen
Manufacturer Code:MA537686
Catalog # PIMA537686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1565CT164.36.80 |
Immunogen | purified recombinant protein of human RAB11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 24KG; 24KG RAB 11A member oncogene family RAB11 rab-11 rab11 GTP-binding protein Rab11a RAB11A member RAS oncogene family ras-related protein Rab-11A tubulovesicle-associated protein YL8; RAB 11A, member oncogene family; Rab-11; Ras-related protein Rab-11A; YL8
Gene Aliases: RAB11; RAB11A; YL8
UniProt ID: (Human) P62491, (Rat) P62494, (Mouse) P62492
Entrez Gene ID: (Human) 8766, (Rat) 81830, (Mouse) 53869
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.