Invitrogen
Manufacturer Code:PA529692
Catalog # PIPA529692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 34 and 166 of Human RAB11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAB 11A, member oncogene family; Rab-11; Ras-related protein Rab-11A; YL8; YL8 RAB 11A member oncogene family rab-11 ras-related protein Rab-11A
Gene Aliases: RAB11; RAB11A; YL8
UniProt ID: (Human) P62491
Entrez Gene ID: (Human) 8766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.