Invitrogen
Manufacturer Code:PA579901
Catalog # PIPA579901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human RAB10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein RAB10; ras-related GTP-binding protein; Ras-related protein Rab-10; Ras-related protein Rab-10 RAB10; ras-related protein rab10
Gene Aliases: Ac1075; AW107754; RAB10
UniProt ID: (Human) P61026, (Mouse) P61027
Entrez Gene ID: (Human) 10890, (Mouse) 19325, (Rat) 50993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.