Invitrogen
Manufacturer Code:PA5102855
Catalog # PIPA5102855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Rab10 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ac1075 AW107754 GTP-binding protein RAB10 RAB10 RAB10 member RAS oncogene family ras-related GTP-binding protein ras-related protein rab10 ras-related protein Rab-10; GTP-binding protein RAB10; ras-related GTP-binding protein; Ras-related protein Rab-10; ras-related protein rab10
Gene Aliases: Ac1075; RAB10
UniProt ID: (Human) P61026
Entrez Gene ID: (Human) 10890, (Rat) 50993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.