Novus Biologicals
Manufacturer Code:NBP293110100UL
Catalog # NB028109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human RAB10 (NP_057215.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein RAB10; GTP-binding protein RAB10 RAB10 member RAS oncogene family ras-related GTP-binding protein ras-related protein Rab-10; ras-related GTP-binding protein; Ras-related protein Rab-10
Gene Aliases: RAB10
UniProt ID: (Human) P61026
Entrez Gene ID: (Human) 10890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.