R&D Systems
Manufacturer Code:MAB3266
Catalog # MAB3266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 879709 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human R-Spondin 2 Met1-Gly205 Accession # Q6UXX9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Cristin 2 CRISTIN2 hRspo2 MGC35555 MGC43342 Roof plate-specific spondin-2 RSPO2 RSpondin 2 R-spondin 2 homolog (Xenopus laevis) R-spondin-2; hRspo2; R-spondin 2 homolog; R-spondin-2; Roof plate-specific spondin-2
Gene Aliases: CRISTIN2; RSPO2; UNQ9384/PRO34209
UniProt ID: (Human) Q6UXX9
Entrez Gene ID: (Human) 340419
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.