Invitrogen
Manufacturer Code:PA569562
Catalog # PIPA569562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of QTRTD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Queuine tRNA-ribosyltransferase accessory subunit 2; queuine tRNA-ribosyltransferase domain containing 1; Queuine tRNA-ribosyltransferase domain-containing protein 1; queuine tRNA-ribosyltransferase domain-containing protein 1 queuine tRNA-ribosyltransferase subunit QTRTD1; queuine tRNA-ribosyltransferase subunit QTRTD1
Gene Aliases: 3110012M05Rik; 4930470H18Rik; AI648807; QTRT2; QTRTD1
UniProt ID: (Human) Q9H974, (Mouse) B8ZXI1
Entrez Gene ID: (Dog) 487983, (Human) 79691, (Pig) 100514190, (Bovine) 508374, (Rabbit) 100346639, (Horse) 100071316, (Guinea pig) 100715795, (Mouse) 106248, (Rat) 288364, (Zebrafish) 402798
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.