Invitrogen
Manufacturer Code:PA543257
Catalog # PIPA543257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human QTRT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Guanine insertion enzyme; queuine tRNA-ribosyltransferase; queuine tRNA-ribosyltransferase 1; queuine tRNA-ribosyltransferase 1 (tRNA-guanine transglycosylase); Queuine tRNA-ribosyltransferase 1 FP3235 TGT TGUT; Queuine tRNA-ribosyltransferase catalytic subunit 1; TGT, 43-KD subunit; TGT, catalytic subunit; tRNA-guanine transglycosylase
Gene Aliases: 2610028E17Rik; FP3235; QTRT1; TGT; TGUT; zgc:66378
UniProt ID: (Human) Q9BXR0, (Mouse) Q9JMA2, (Rat) Q4QR99, (Zebrafish) Q7SYK1
Entrez Gene ID: (Bovine) 505497, (Rabbit) 100348707, (Human) 81890, (Dog) 476711, (Horse) 100057666, (Mouse) 60507, (Rat) 64016, (Guinea pig) 100723403, (Zebrafish) 393985
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.