Invitrogen
Manufacturer Code:PA531270
Catalog # PIPA531270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 22 and 340 of Human QTRT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FP3235 TGT guanine insertion enzyme queuine tRNA-ribosyltransferase tRNA-guanine transglycosylase; Guanine insertion enzyme; queuine tRNA-ribosyltransferase; queuine tRNA-ribosyltransferase 1; Queuine tRNA-ribosyltransferase catalytic subunit 1; TGT, 43-KD subunit; TGT, catalytic subunit; tRNA-guanine transglycosylase
Gene Aliases: 2610028E17Rik; FP3235; QTRT1; TGT; TGUT
UniProt ID: (Human) Q9BXR0, (Mouse) Q9JMA2
Entrez Gene ID: (Human) 81890, (Mouse) 60507
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.