Invitrogen
Manufacturer Code:PA570797
Catalog # PIPA570797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse QSOX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuroblastoma-derived sulfhydryl oxidase; QSCN6L1 SOXN neuroblastoma-derived sulfhydryl oxidase quiescin Q6-like 1 quiescin Q6-like protein 1 sulfhydryl oxidase 2; quiescin Q6 sulfhydryl oxidase 2; quiescin Q6-like 1; Quiescin Q6-like protein 1; Sulfhydryl oxidase 2; thiol oxidase 2
Gene Aliases: BC030934; QSCN6L1; QSOX2; SOXN
UniProt ID: (Human) Q6ZRP7, (Mouse) Q3TMX7
Entrez Gene ID: (Dog) 607571, (Human) 169714, (Rat) 681023, (Mouse) 227638, (Guinea pig) 100722911
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.