Invitrogen
Manufacturer Code:PA534150
Catalog # PIPA534150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from internal region of human QSOX2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuroblastoma-derived sulfhydryl oxidase; QSOX2 Quiescin Q6-like protein 1 SOXN Sulfhydryl oxidase 2 QSCN6L1 Quiescin Q6-like 1; quiescin Q6 sulfhydryl oxidase 2; quiescin Q6-like 1; Quiescin Q6-like protein 1; Sulfhydryl oxidase 2; thiol oxidase 2
Gene Aliases: QSCN6L1; QSOX2; SOXN
UniProt ID: (Human) Q6ZRP7
Entrez Gene ID: (Human) 169714
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.