Invitrogen
Manufacturer Code:PA538009
Catalog # PIPA538009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 17 amino acid peptide near the center of human QSOX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hQSOX; mSOx; Quiescin Q6; quiescin Q6 sulfhydryl oxidase 1; Quiescin Q6 sulfhydryl oxidase 1 Sulfhydryl oxidase 1 Q6 QSCN6; Skin sulfhydryl oxidase; Sulfhydryl oxidase 1; testis tissue sperm-binding protein Li 62n; thiol oxidase 1
Gene Aliases: 1300003H02Rik; Q6; QSCN6; QSOX; QSOX1; SOx; UNQ2520/PRO6013
UniProt ID: (Human) O00391, (Mouse) Q8BND5
Entrez Gene ID: (Human) 5768, (Mouse) 104009
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.