Invitrogen
Manufacturer Code:PA521578
Catalog # PIPA521578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 231 and 617 of QSOX1 (Uniprot IDNo.O00391) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hQSOX; Quiescin Q6; quiescin Q6 sulfhydryl oxidase 1; RP11-502H18.3 Q6 QSCN6 hQSOX sulfhydryl oxidase 1; Sulfhydryl oxidase 1; testis tissue sperm-binding protein Li 62n; thiol oxidase 1
Gene Aliases: Q6; QSCN6; QSOX1; UNQ2520/PRO6013
UniProt ID: (Human) O00391
Entrez Gene ID: (Human) 5768
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.