Invitrogen
Manufacturer Code:PA556199
Catalog # PIPA556199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human QRSL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glu-AdT subunit A; glu-AdT subunit A {ECO:0000255|HAMAP-Rule:MF_03150}; Glutaminyl-tRNA synthase-like protein 1; glutaminyl-tRNA synthase-like protein 1 {ECO:0000255|HAMAP-Rule:MF_03150}; glutamyl-tRNA(Gln) amidotransferase subunit A homolog; Glutamyl-tRNA(Gln) amidotransferase subunit A mitochondrial Glu-AdT subunit A 6357 Glutaminyl-tRNA synthase-like protein 1 QRSL1; Glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03150}; glutamyl-tRNA(Gln) amidotransferase, subunit A; QRSL1
Gene Aliases: 2700038P16Rik; C80053; GatA; QRSL1
UniProt ID: (Human) Q9H0R6, (Rat) Q5FWT5, (Mouse) Q9CZN8
Entrez Gene ID: (Human) 55278, (Rat) 309911, (Mouse) 76563
Molecular Function: hydrolase ligase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.