Invitrogen
Manufacturer Code:PA558052
Catalog # PIPA558052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human QRICH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ20259 MGC131838 Glutamine-rich Protein 1; glutamine-rich 1; Glutamine-rich protein 1
Gene Aliases: 2610028H07Rik; QRICH1; RGD1311095
UniProt ID: (Human) Q2TAL8, (Mouse) Q3UA37
Entrez Gene ID: (Human) 54870, (Rat) 301004, (Mouse) 69232
Molecular Function:
enzyme modulator
kinase inhibitor
kinase modulator
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.