Invitrogen
Manufacturer Code:PA559303
Catalog # PIPA559303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human QPCTL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutaminyl cyclase-like; Glutaminyl-peptide cyclotransferase-like protein; Glutaminyl-peptide cyclotransferase-like protein Golgi-resident glutaminyl-peptide cyclotransferase isoQC gQC QPCTL; Golgi-resident glutaminyl-peptide cyclotransferase; gQC; isoQC
Gene Aliases: gQC; QPCTL
UniProt ID: (Human) Q9NXS2
Entrez Gene ID: (Human) 54814
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.