Novus Biologicals
Manufacturer Code:NBP242196B
Catalog # NBP242196B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S147-6 |
Immunogen | Fusion protein amino acids 1-341 (full-length) of human QKI-5. Mouse: 100% identity (341/341 amino acids identical). Rat: 99% identity (339/341 amino acids identical) >90% identity with QKI-6 QKI-7 and QKI-7b. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp586I0923 HKQ homolog of mouse quaking QKI (KH domain RNA binding protein) Hqk hqkI protein quaking QK QK1 QK3 quaking homolog KH domain RNA binding (mouse) RNA binding protein HQK; homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; Protein quaking; QKI, KH domain containing, RNA binding; quaking homolog, KH domain RNA binding; RNA binding protein HQK
Gene Aliases: HKQ; Hqk; hqkI; QK; QK1; QK3; QKI
UniProt ID: (Human) Q96PU8
Entrez Gene ID: (Human) 9444
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.