Novus Biologicals
Manufacturer Code:NB300239
Catalog # NB300239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide mapping to a region between residues 50 and 100 of isoform 5 of human quaking homolog using the numbering given in entry AAH19917.1 (GeneID 9444). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586I0923 HKQ homolog of mouse quaking QKI (KH domain RNA binding protein) Hqk hqkI protein quaking QK QK1 QK3 quaking homolog KH domain RNA binding (mouse) RNA binding protein HQK; homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; Protein quaking; QKI, KH domain containing, RNA binding; quaking homolog, KH domain RNA binding; RNA binding protein HQK
Gene Aliases: HKQ; Hqk; hqkI; QK; QK1; QK3; QKI
UniProt ID: (Human) Q96PU8
Entrez Gene ID: (Human) 9444
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.