Invitrogen
Manufacturer Code:PA553930
Catalog # PIPA553930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human QKI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; lethal, Chr 17, U Wisconsin 1; MqkI; Protein quaking; protein quaking-like; QKI quaking homolog KH domain RNA binding DKFZp586I0923 Hqk QK QK3 RNA binding protein HQK homolog of mouse quaking QKI (KH domain RNA binding protein) quaking homolog KH domain RNA binding; QKI, KH domain containing, RNA binding; QKI-5 protein; quaking homolog, KH domain RNA binding; RNA binding protein HQK
Gene Aliases: 1110003F05Rik; 1500005P18; HKQ; Hqk; hqkI; l(17)-1Wis; l17Wis1; QK; QK1; QK3; Qka1; QKI
UniProt ID: (Human) Q96PU8, (Mouse) Q9R2A8
Entrez Gene ID: (Human) 9444, (Mouse) 19317, (Rat) 100910212
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.