Invitrogen
Manufacturer Code:PA528041
Catalog # PIPA528041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 244 of Human QDPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6,7-dihydropteridine reductase; DHPR PKU2 SDR33C1 67-dihydropteridine reductase HDHPR dihydropteridine reductase short chain dehydrogenase/reductase family 33C member 1; Dihydropteridine reductase; HDHPR; Quinoid dihydropteridine reductase; Short chain dehydrogenase/reductase family 33C member 1; short chain dehydrogenase/reductase family 33C, member 1; testis secretory sperm-binding protein Li 236P
Gene Aliases: 2610008L04Rik; D5Ertd371e; DHPR; PKU2; QDPR; SDR33C1
UniProt ID: (Human) P09417, (Mouse) Q8BVI4
Entrez Gene ID: (Human) 5860, (Mouse) 110391
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.