R&D Systems
Manufacturer Code:AF8038
Catalog # AF8038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human QDPR Ala2-Phe244 Accession # P09417 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 6,7-dihydropteridine reductase; dhpr DHPR member 1 HDHPR PKU2 quinoid dihydropteridine reductaseFLJ42391 SDR33C1; Dihydropteridine reductase; HDHPR; Quinoid dihydropteridine reductase; Short chain dehydrogenase/reductase family 33C member 1; short chain dehydrogenase/reductase family 33C, member 1; testis secretory sperm-binding protein Li 236P
Gene Aliases: DHPR; PKU2; QDPR; SDR33C1
UniProt ID: (Human) P09417
Entrez Gene ID: (Human) 5860
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.