Novus Biologicals
Manufacturer Code:NBP177180
Catalog # NBP177180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human FREM1. The immunogen is located within the last 50 amino acids of FREM1. Amino Acid Squence: TNGRRGPSPRSKLGK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BNAR C9orf143chromosome 9 open reading frame 154 C9orf145C9orf154extracellular matrix protein QBRICK DKFZp686M16108 FLJ25461 FRAS1 related extracellular matrix 1 FRAS1-related extracellular matrix protein 1 Protein QBRICK TILRR; extracellular matrix protein QBRICK; FRAS1-related extracellular matrix protein 1; Protein QBRICK
Gene Aliases: BNAR; C9orf143; C9orf145; C9orf154; FREM1; MOTA; TILRR; TRIGNO2
UniProt ID: (Human) Q5H8C1
Entrez Gene ID: (Human) 158326
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.