Novus Biologicals
Manufacturer Code:NBP298572JF549
Catalog # NB054354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human QARS fragment. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.1.1 EC 6.1.1.18 GLNRS glutamine tRNA ligase Glutamine--tRNA ligase glutamine-tRNA synthetase glutaminyl-tRNA synthetase PRO2195; GlnRS; Glutamine--tRNA ligase; glutamine-tRNA synthetase; Glutaminyl-tRNA synthetase
Gene Aliases: GLNRS; MSCCA; PRO2195; QARS; QARS1
UniProt ID: (Human) P47897
Entrez Gene ID: (Human) 5859
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.