Novus Biologicals
Manufacturer Code:NBP226649SS
Catalog # NBP226649SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6A8.6F10.1A6 |
Immunogen | Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells. The antibody reacts with the Qa-1b Class Ib molecule. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: H-2 class I histocompatibility antigen, D-37 alpha chain; H2-Q1 H2-T23; MHC class I antigen Qa1; MHC classIb Qa1
Gene Aliases: 37b; 37c; H-2T23; H2-Qa1; H2-T23; Qa-1; Qa-1(b); Qa1; Qed-1; T18c; T18c(37); T23b; T23d
UniProt ID: (Mouse) P06339
Entrez Gene ID: (Mouse) 15040
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.