Novus Biologicals
Manufacturer Code:NBP149536V
Catalog # NBP149536V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Partial recombinant protein made to an internal region of the human Pyruvate Carboxylase protein (within residues 930-1050). [Swiss-Prot P11498] |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: EC 6.4.1 EC 6.4.1.1 PCBPyruvic carboxylase pyruvate carboxylase pyruvate carboxylase mitochondrial; PCB; Pyruvate carboxylase, mitochondrial; Pyruvic carboxylase
Gene Aliases: PC; PCB
UniProt ID: (Human) P11498
Entrez Gene ID: (Human) 5091
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.