Novus Biologicals
Manufacturer Code:NBP146171
Catalog # NBP146171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 1 and 50 of human Pygopus 2 using the numbering given in entry NP_612157.1 (GeneID 90780). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1190004M21Rik FLJ33226 pygopus 2 pygopus homolog 2 pygopus homolog 2 (Drosophila); pygopus 2; Pygopus homolog 2
Gene Aliases: 1190004M21Rik; PP7910; PYGO2
UniProt ID: (Human) Q9BRQ0
Entrez Gene ID: (Human) 90780
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.