Novus Biologicals
Manufacturer Code:NBP288103
Catalog # NB035725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human Puromycin-sensitive aminopeptidase/NPEPPS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAP-S; AAP-S aminopeptidase puromycin sensitive Cytosol alanyl aminopeptidase EC 3.4.11.2 EC 3.4.11.3 metalloproteinase MP100 MP100EC 3.4.11 PSAEC 3.4.11.14 puromycin-sensitive aminopeptidase; Cytosol alanyl aminopeptidase; metalloproteinase MP100; PSA; Puromycin-sensitive aminopeptidase
Gene Aliases: AAP-S; MP100; NPEPPS; PSA
UniProt ID: (Human) P55786
Entrez Gene ID: (Human) 9520
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.