R&D Systems
Manufacturer Code:MAB6486SP
Catalog # MAB6486SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 773511 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Purine Nucleoside Phosphorylase/PNP Met1-Ser289 Accession # P00491 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.2.1 FLJ94043 FLJ97288 Inosine phosphorylase MGC117396 MGC125915 MGC125916 NPFLJ97312 nucleoside phosphorylase PRO1837 PUNP purine nucleoside phosphorylase purine-nucleoside:orthophosphate ribosyltransferase; epididymis secretory sperm binding protein Li 156an; HEL-S-156an; Inosine phosphorylase; Inosine-guanosine phosphorylase; PNP; Purine nucleoside phosphorylase; purine-nucleoside:orthophosphate ribosyltransferase
Gene Aliases: NP; PNP; PRO1837; PUNP
UniProt ID: (Human) P00491
Entrez Gene ID: (Human) 4860
Molecular Function:
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.