Abnova
Manufacturer Code:MAB2149
Catalog # 89-110-848
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP21 |
Immunogen | A synthetic peptide corresponding to C-terminus of rat Ptgs2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit monoclonal antibody raised against synthetic peptide of Ptgs2.
For Research Use Only
Protein Aliases: COX-2; Cox2; cyclooxygenase 2; Cyclooxygenase-2; PGH synthase 2; PGHS-2; PHS II; Prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; Prostaglandin-endoperoxide synthase 2
Gene Aliases: COX-2; Cox2; Ptgs2
UniProt ID: (Rat) P35355
Entrez Gene ID: (Rat) 29527
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.