Abnova
Manufacturer Code:MAB6624
Catalog # 89-112-958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6G6 |
Immunogen | Recombinant protein corresponding to rat Psd. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant Psd.
For Research Use Only
Protein Aliases: EFA6; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ARF6; Exchange factor for ARF6 A; PH and SEC7 domain-containing protein 1; Pleckstrin homology and SEC7 domain-containing protein 1
Gene Aliases: EFA6; Psd; Psd1
UniProt ID: (Rat) Q9ESQ7
Entrez Gene ID: (Rat) 171381
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.