Novus Biologicals
Manufacturer Code:NBP130045PE
Catalog # NBP130045PE
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5G10 |
Immunogen | The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786] |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Homeobox prospero-like protein PROX1; Homeobox prospero-like protein PROX1 prospero homeobox 1 prospero homeobox protein 1 prospero-related homeobox 1 PROX-1; Prospero homeobox protein 1; prospero-related homeobox 1; PROX-1
Gene Aliases: PROX1
UniProt ID: (Human) Q92786
Entrez Gene ID: (Human) 5629
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.