R&D Systems
Manufacturer Code:AF6729
Catalog # AF6729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin-15 Gln27-Ala1376 Accession # Q96QU1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: autosomal recessive 23 cadherin-related family member 15 CDHR15 DFNB23 DKFZp667A1711 PCDH15 protocadherin-15 Protocadherin15 protocadherin-related 15 USH1F; cadherin-related family member 15; Protocadherin-15; protocadherin-related 15
Gene Aliases: CDHR15; DFNB23; PCDH15; USH1F
UniProt ID: (Human) Q96QU1
Entrez Gene ID: (Human) 65217
Molecular Function:
cadherin
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.