Invitrogen
Manufacturer Code:MA143034
Catalog # 01-674-555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 9013 |
Immunogen | Mouse prothrombin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation peptide fragment 1; Activation peptide fragment 2; Coagulation factor II; Prothrombin; Thrombin heavy chain; Thrombin light chain
Gene Aliases: Cf-2; Cf2; F2; FII
UniProt ID: (Mouse) P19221
Entrez Gene ID: (Mouse) 14061
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.