EMD Millipore
Manufacturer Code:MABF285
Catalog # MABF285MI
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 29J3-62 |
Immunogen | Purified human prothrombin (PT) and Dioleoylphosphatidylserine (DOPS) complex. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | &minus20°C for one year from date of shipment Upon first thaw,and prior to removing the cap,centrifuge the vial and gently mix the solution Aliquot into microcentrifuge tubes and store at &minus20°C Avoid repeated freeze/thaw cycles |
Mouse Monoclonal Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.