Invitrogen
Manufacturer Code:PA584070
Catalog # PIPA584070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human F2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activation peptide fragment 1; Activation peptide fragment 2; Coagulation factor II; coagulation factor II (thrombin); prepro-coagulation factor II; Prothrombin; prothrombin B-chain; serine protease; Thrombin heavy chain; Thrombin light chain
Gene Aliases: F2; PT; RPRGL2; THPH1
UniProt ID: (Human) P00734
Entrez Gene ID: (Human) 2147
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.