Invitrogen
Manufacturer Code:PA569150
Catalog # PIPA569150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat Prothrombin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; coagulation factor 2; Coagulation factor II; coagulation factor II (thrombin); Coagulation factor II F2; kringle-2 domain; fragment-2; prepro-coagulation factor II; preprothrombin; Prothrombin; prothrombin B-chain; serine protease; thrombin; Thrombin heavy chain; Thrombin light chain
Gene Aliases: Cf-2; Cf2; F2; FII; PT; RPRGL2; THPH1; THROMBIN
UniProt ID: (Pig) Q19AZ8, (Mouse) P19221, (Human) P00734, (Bovine) P00735
Entrez Gene ID: (Pig) 100144442, (Mouse) 14061, (Sheep) 100307041, (Rat) 29251, (Guinea pig) 100725988, (Human) 2147, (Bovine) 280685, (Rabbit) 100009146, (Horse) 100057521, (Dog) 100856539
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.