R&D Systems
Manufacturer Code:MAB64371
Catalog # MAB64371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 737512 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Protein O-Glucosyltransferase 1/POGLUT Arg24-Leu388 Accession # Q8NBL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 9630046K23Rik; 9630046K23Rik C3orf9 CAP10-like protein 46 kDa chromosome 3 open reading frame 9 CLP46 EC 2.4.1.- hCLP46CAP10-like 46 kDa protein KDELC family like 1 KDELCL1 KTEL (Lys-Tyr-Glu-Leu) containing 1 KTEL motif-containing protein 1 KTELC1 MDS010 MDSRP MDSRPKTELC1 MGC32995 Myelodysplastic syndromes relative protein POGLUT1 protein O-glucosyltransferase 1 Rumi x 010 protein; CAP10-like 46 kDa protein; hCLP46; hRumi; KDELC family like 1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; x 010 protein
Gene Aliases: C3orf9; CLP46; hCLP46; KDELCL1; KTELC1; MDS010; MDSRP; POGLUT1; Rumi; UNQ490/PRO1006
UniProt ID: (Human) Q8NBL1
Entrez Gene ID: (Human) 56983
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.