Invitrogen
Manufacturer Code:PA5111439
Catalog # PIPA5111439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human Protein Kinase D2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APKD2 Autosomal dominant polycystic kidney disease type II protein DKFZP586E0820 EPK2 HSPC187 nPKC-D1 nPKC-D2 nPKC-mu nPKC-nu PC2 Pc-2 PKC mu PKC nu PKC u PKC v PKCmu PKCnu PKC-nu PKCu PKC-u PKCv PKC-v PKD PKD1 PKD2 PKD3 PKD4 Polycystic kidney disease 2 protein Polycystin 2 Polycystin-2 Polycystwin PRKCM PRKCN PRKD1 PRKD2 PRKD3 Protein kinase D Protein kinase D2 Protein kinase EPK2 R48321 Serine/threonine-protein kinase D2 Transient receptor potential cation channel subfamily P member 2 TRPP2; Autosomal dominant polycystic kidney disease type II protein; nPKC-D2; PC2; polycystic kidney disease 2 (autosomal dominant); Polycystic kidney disease 2 protein; Polycystin-2; Polycystwin; R48321; Serine/threonine-protein kinase D2; Transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2
Gene Aliases: APKD2; HSPC187; nPKC-D2; Pc-2; PC2; PKD2; PKD4; PRKD2; TRPP2
UniProt ID: (Human) Q13563, (Human) Q9BZL6
Entrez Gene ID: (Human) 5311, (Human) 25865
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.