Novus Biologicals
Manufacturer Code:NBP289676AF405
Catalog # NB099636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 16 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Protein Disulfide Isomerase/P4HB (NP_000909.2 Met 1-Lys 505). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Cellular thyroid hormone-binding protein; Cellular thyroid hormone-binding protein collagen prolyl 4-hydroxylase beta DSI ERBA2L GIT glutathione-insulin transhydrogenase P4Hbeta p55 PDIA1procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) betapolypeptide (protein disulfide isomerase-associated 1) PDIEC 5.3.4.1 PHDB PO4DB PO4HB procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) betapolypeptide PROHBprocollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) betapolypeptide (protein disulfide isomerase thyroid hormone binding protein p55) Prolyl 4-hydroxylase subunit beta prolyl 4-hydroxylase beta polypeptide protein disulfide isomerase family A member 1 protein disulfide isomerase/oxidoreductase protein disulfide isomerase-associated 1 protein disulfide-isomerase protocollagen hydroxylase thyroid hormone-binding protein p55; collagen prolyl 4-hydroxylase beta; glutathione-insulin transhydrogenase; p55; PDI; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), beta polypeptide; Prolyl 4-hydroxylase subunit beta; prolyl 4-hydroxylase, beta polypeptide; protein disulfide isomerase family A, member 1; protein disulfide isomerase-associated 1; protein disulfide isomerase/oxidoreductase; Protein disulfide-isomerase; protocollagen hydroxylase; testicular secretory protein Li 32; thyroid hormone-binding protein p55
Gene Aliases: CLCRP1; DSI; ERBA2L; GIT; P4HB; P4Hbeta; PDI; PDIA1; PHDB; PO4DB; PO4HB; PROHB
UniProt ID: (Human) P07237
Entrez Gene ID: (Human) 5034
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.