R&D Systems
Manufacturer Code:BAF4236
Catalog # BAF4236
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Protein Disulfide Isomerase/P4HB Asp18-Lys505 Accession # P07237 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Proline 4-Hydroxylase, Beta Polypeptide
For Research Use Only
Protein Aliases: Cellular thyroid hormone-binding protein; Cellular thyroid hormone-binding protein collagen prolyl 4-hydroxylase beta DSI ERBA2L GIT glutathione-insulin transhydrogenase P4Hbeta p55 PDI PDIA1 PDIA1procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) betapolypeptide (protein disulfide isomerase-associated 1) PDIEC 5.3.4.1 PHDB PO4DB PO4HB procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) betapolypeptide PROHB PROHBprocollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) betapolypeptide (protein disulfide isomerase thyroid hormone binding protein p55) Prolyl 4-hydroxylase subunit beta prolyl 4-hydroxylase beta polypeptide protein disulfide isomerase family A member 1 protein disulfide isomerase/oxidoreductase protein disulfide isomerase-associated 1 protein disulfide-isomerase protocollagen hydroxylase Thbp thyroid hormone-binding protein p55; collagen prolyl 4-hydroxylase beta; glutathione-insulin transhydrogenase; p55; PDI; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), beta polypeptide; Prolyl 4-hydroxylase subunit beta; prolyl 4-hydroxylase, beta polypeptide; protein disulfide isomerase family A, member 1; protein disulfide isomerase-associated 1; protein disulfide isomerase/oxidoreductase; Protein disulfide-isomerase; protocollagen hydroxylase; testicular secretory protein Li 32; thyroid hormone-binding protein p55
Gene Aliases: CLCRP1; DSI; ERBA2L; GIT; P4HB; P4Hbeta; PDI; PDIA1; PHDB; PO4DB; PO4HB; PROHB
UniProt ID: (Human) P07237
Entrez Gene ID: (Human) 5034
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.