Invitrogen
Manufacturer Code:MA536220
Catalog # PIMA536220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | RM426 |
Immunogen | A peptide corresponding to the C-terminus of human Prostein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2210413P12Rik AU023994 AU043764 IPCA-2 IPCA6 IPCA-6 IPCA-8 PCANAP2 Pcanap6 PCANAP8 prostate cancer associated protein 2 prostate cancer associated protein 6 prostate cancer associated protein 8 prostate cancer-associated gene 2 prostate cancer-associated gene 6 prostate cancer-associated gene 8 prostate cancer-associated protein 6 Prostein PRST RGD1309764 SLC45A3 solute carrier family 45 member 3 solute carrier family 45 member 3; prostate cancer associated protein 2; prostate cancer associated protein 6; prostate cancer associated protein 8; prostate cancer-associated gene 2; prostate cancer-associated gene 6; prostate cancer-associated gene 8; Prostate cancer-associated protein 6; Prostein; Solute carrier family 45 member 3; solute carrier family 45, member 3
Gene Aliases: IPCA-2; IPCA-6; IPCA-8; IPCA6; PCANAP2; PCANAP6; PCANAP8; PRST; SLC45A3
UniProt ID: (Human) Q96JT2
Entrez Gene ID: (Human) 85414
Molecular Function: carbohydrate transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.