R&D Systems
Manufacturer Code:AF6240
Catalog # AF6240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Prostatic Acid Phosphatase/ACPP Lys33-Gln379 Accession # P15309 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-NT; 5'-nucleotidase; Acid phosphatase 3; acid phosphatase prostate ACP3 ACP-3 ACPP EC 3.1.3.2 PAP prostatic acid phosphatase prostatic acid phosphotase; Ecto-5'-nucleotidase; PAP; PAPf39; Prostatic acid phosphatase; prostatic acid phosphotase; Protein tyrosine phosphatase ACP3; Thiamine monophosphatase; TMPase
Gene Aliases: 5'-NT; ACP-3; ACP3; ACPP
UniProt ID: (Human) P15309
Entrez Gene ID: (Human) 55
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.