Invitrogen
Manufacturer Code:MA538108
Catalog # PIMA538108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2124 |
Immunogen | A synthesized peptide derived from human Prostatic Acid Phosphatase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 5'-NT; 5'-NT 5'-nucleotidase A030005E02Rik acid phosphatase prostate ACP3 ACP-3 Acpp Acpp11 EC 3.1.3.2 Ecto-5'-nucleotidase fluoride-resistant acid phosphatase FRAP Lap lysosomal acid phosphatase pap PAPf39 Ppal PPAP prostate acid phosphatase prostatic acid phosphatase prostatic acid phosphatase (rPAP) prostatic acid phosphotase RNACPP11 Thiamine monophosphatase TMPase; 5'-nucleotidase; Acid phosphatase 3; Ecto-5'-nucleotidase; Fluoride-resistant acid phosphatase; FRAP; lysosomal acid phosphatase; PAP; PAPf39; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; Protein tyrosine phosphatase ACP3; Thiamine monophosphatase; TMPase
Gene Aliases: 5'-NT; A030005E02Rik; ACP-3; ACP3; ACPP; Acpp11; FRAP; Lap; PAP; Ppal; RNACPP11
UniProt ID: (Human) P15309, (Mouse) Q8CE08, (Rat) P20646
Entrez Gene ID: (Human) 55, (Mouse) 56318, (Rat) 56780
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.