Invitrogen
Manufacturer Code:MA530059
Catalog # PIMA530059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 202 |
Immunogen | Recombit Mouse Prostatic Acid Phosphatase/ACPP protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 5'-NT; 5'-nucleotidase; Acid phosphatase 3; Ecto-5'-nucleotidase; Fluoride-resistant acid phosphatase; FRAP; lysosomal acid phosphatase; Prostatic acid phosphatase; Prostatic acid phosphatase &lsqbPrecursor] PPAP EC 3.1.3.2 ACPP; Protein tyrosine phosphatase ACP3; Thiamine monophosphatase; TMPase
Gene Aliases: 5'-NT; A030005E02Rik; Acp3; Acpp; FRAP; Lap; PAP; Ppal
UniProt ID: (Mouse) Q8CE08
Entrez Gene ID: (Mouse) 56318
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.