Novus Biologicals
Manufacturer Code:NBP273623JF646
Catalog # N273623J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C3 |
Immunogen | Full length recombinant protein of human Prostaglandin E Synthase 2/PTGES2 (NP_079348) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C9orf15 chromosome 9 open reading frame 15 EC 5.3.99.3 FLJ14038 gamma-interferon-activated transcriptional element-binding factor 1 GATE-binding factor 1 GBF1 membrane-associated prostaglandin E synthase 2 Microsomal prostaglandin E synthase 2 microsomal prostaglandin E synthase-2 mPGES-2 PGES2MGC11289 prostaglandin E synthase 2; gamma-interferon-activated transcriptional element-binding factor 1; GATE-binding factor 1; membrane-associated prostaglandin E synthase 2; Membrane-associated prostaglandin E synthase-2; Microsomal prostaglandin E synthase 2; microsomal prostaglandin E synthase-2; mPGE synthase-2; mPGES-2; Prostaglandin E synthase 2; Prostaglandin E synthase 2 truncated form; Prostaglandin-H(2) E-isomerase
Gene Aliases: C9orf15; GBF-1; GBF1; mPGES-2; PGES2; PTGES2
UniProt ID: (Human) Q9H7Z7
Entrez Gene ID: (Human) 80142
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.