R&D Systems
Manufacturer Code:MAB6018
Catalog # MAB6018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 694009 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Proprotein Convertase 2/PCSK2 Gly110-Asn638 Accession # P16519 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.21 EC 3.4.21.94 KEX2-like endoprotease 2 NEC 2 NEC2 NEC2SPC2 neuroendocrine convertase 2 PC2 PC2NEC-2 PCSK2 Prohormone convertase 2 Proprotein convertase 2 proprotein convertase subtilisin/kexin type 2 SPC2; KEX2-like endoprotease 2; NEC 2; Neuroendocrine convertase 2; PC2; Prohormone convertase 2; Proprotein convertase 2
Gene Aliases: NEC 2; NEC-2; NEC2; PC2; PCSK2; SPC2
UniProt ID: (Human) P16519
Entrez Gene ID: (Human) 5126
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.