R&D Systems
Manufacturer Code:MAB2024
Catalog # MAB2024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 244029 |
Immunogen | NS0 mouse myeloma cell line transfected with human Prominin 2 Ala27-Leu834 Accession # AAM10541 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hPROML2; hPROML2 MGC138714 PROM2 PROM-2 prominin 2 prominin-2 Prominin-like protein 2 prominin-related protein PROML2; PROM-2; Prominin-2; Prominin-like protein 2; prominin-related protein
Gene Aliases: PROM2; PROML2; UNQ2521/PRO6014
UniProt ID: (Human) Q8N271
Entrez Gene ID: (Human) 150696
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.