Invitrogen
Manufacturer Code:PA532995
Catalog # PIPA532995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from N-terminus of human PROM2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPROML2; PROM-2; Prominin-2; Prominin-like protein 2; prominin-related protein; PROML2 hPROML2 prominin-2 prominin-like protein 2 prominin-related protein
Gene Aliases: PROM2; PROML2; UNQ2521/PRO6014
UniProt ID: (Human) Q8N271
Entrez Gene ID: (Human) 150696
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.