Novus Biologicals
Manufacturer Code:NBP29330520UL
Catalog # NB028496
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 578-727 of human PREPL (NP_006027.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.21 EC 3.4.21.- EC 3.4.21.83 KIAA0436FLJ16627 prolyl endopeptidase-like Prolylendopeptidase-like putative prolyl oligopeptidase; Prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase
Gene Aliases: KIAA0436; PREPL
UniProt ID: (Human) Q4J6C6
Entrez Gene ID: (Human) 9581
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.