Novus Biologicals
Manufacturer Code:NBP289378100UL
Catalog # NB020239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 313 |
Immunogen | Recombinant Human Prolactin R Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: delta 4-delta 7/11 truncated prolactin receptor delta 4-SF1b truncated prolactin receptor hPRL receptor hPRLrI PRL-R prolactin receptor prolactin receptor delta 7/11 secreted prolactin binding protein; hPRL receptor; PRL-R; Prolactin receptor; secreted prolactin binding protein
Gene Aliases: HPRL; hPRLrI; MFAB; PRLR
UniProt ID: (Human) P16471
Entrez Gene ID: (Human) 5618
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.